Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178372.2 | NP_848459.1 | 1 - 2 | 173 | 63 | |
GenBank mRNA | AK081986.1 | - | 1 - 2 | 175 | 63 | |
AY261775.1 | - | 1 - 2 | 173 | 63 | ||
BC109366.2 | - | 1 - 2 | 147 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |