Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178398.4 | NP_848485.1 | 5 - 6 | 698 | 72 | |
GenBank mRNA | AK043857.1 | - | 4 - 5 | 1507 | 72 | |
AK155113.1 | - | 5 - 6 | 728 | 72 | ||
AK169051.1 | - | 5 - 6 | 672 | 72 | ||
AK172030.1 | - | 5 - 6 | 674 | 72 | ||
BC044894.1 | - | 5 - 6 | 723 | 4235237 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |