Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178778.3 | NP_848893.2 | 13 - 14 | 1331 | 94 | |
GenBank mRNA | AK044772.1 | - | 13 - 14 | 1327 | 94 | |
AK082408.1 | - | 14 - 15 | 1433 | 94 | ||
BC132485.1 | - | 13 - 14 | 1306 | 40130762 | 94 | |
BC144986.1 | - | 14 - 15 | 1407 | 94 | ||
BC144987.1 | - | 13 - 14 | 1382 | 94 | ||
BC144989.1 | - | 13 - 14 | 1305 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |