Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178785.3 | NP_848900.2 | 12 - 13 | 2373 | 65 | |
GenBank mRNA | AK030797.1 | - | 12 - 13 | 2376 | 65 | |
AK040573.1 | - | 12 - 13 | 2361 | 65 | ||
AK041479.1 | - | 13 - 14 | 2194 | 65 | ||
AK088449.1 | - | 12 - 13 | 2335 | 65 | ||
AK088987.1 | - | 12 - 13 | 2351 | 65 | ||
AK131128.1 | - | 14 - 15 | 2280 | 65 | ||
BC132341.1 | - | 14 - 15 | 2279 | 40130618 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |