Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181402.3 | NP_852067.1 | 2 - 3 | 219 | 104 | |
GenBank mRNA | AK133697.1 | - | 2 - 3 | 267 | 104 | |
BC040269.1 | - | 2 - 3 | 219 | 4035469 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |