Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2094016 |
AK162320.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2094016 |
BC147415.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2094016 |
BC147416.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2094016 |
AK033669.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2094016 |
AK050272.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2094019 |
AK050272.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2094019 |
NM_177016.3 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
484542 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
384096 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
876297 |
AK051349.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
876297 |
AK150279.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
876297 |
BC025514.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
876297 |
BC026022.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
876297 |
BC028824.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
876297 |
BC034836.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
876297 |
NM_178644.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
453216 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
701258 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
943590 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
223020 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
327668 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
342762 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
913245 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
100537 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
96299 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P101382 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P154997 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2094016 |
AK162320.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2094016 |
BC147415.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2094016 |
BC147416.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2094016 |
AK033669.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2094016 |
AK050272.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2094019 |
AK050272.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2094019 |
NM_177016.3 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884274 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884292 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884255 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884278 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884297 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884264 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884259 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884267 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884276 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884295 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884261 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884269 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884300 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884272 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884273 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884284 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884296 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884288 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884286 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884285 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884270 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884254 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884256 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884280 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884265 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884281 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884277 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884260 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884266 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28884291 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P154997 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07810927 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P101382 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P477785 |
AK162320.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P477785 |
BC147415.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P477785 |
BC147416.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P477785 |
AK033669.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P477785 |
AK050272.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P140862 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P126769 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P101382 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P154997 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10874475 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10874473 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10874477 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10852008 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10852007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884326 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884303 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884325 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884228 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884253 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884168 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884259 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884153 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884302 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884241 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884305 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884225 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884293 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884172 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884156 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884280 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884274 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884278 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884090 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884243 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884170 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884208 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884284 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884164 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884261 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884235 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884193 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884165 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884286 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884295 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884308 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884097 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884327 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884252 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884250 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884255 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884222 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884217 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884320 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884186 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884191 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884112 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884213 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884089 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884102 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884151 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884246 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884296 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884265 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884205 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884232 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884270 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884239 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884173 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884100 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884181 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884174 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884154 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884226 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884233 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884155 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884171 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884160 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884220 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884224 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884215 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884152 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884197 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884251 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884204 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884269 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884237 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884159 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884184 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884310 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884098 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884323 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884179 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884288 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884190 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884231 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884300 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884177 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884249 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884195 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884256 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884162 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884166 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28884175 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884286 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884251 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884231 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884278 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10884929 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884303 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884302 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884274 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884225 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884284 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10694260 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884239 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10533737 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10695972 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10874474 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884265 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884273 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884259 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884255 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884291 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884246 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884233 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10858715 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884293 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884281 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884245 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884269 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884250 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11013568 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884237 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884244 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10852008 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884252 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884292 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884295 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884228 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10874470 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884288 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884241 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884270 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884266 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884235 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884296 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884276 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884256 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884308 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884289 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884306 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884305 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884253 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884260 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884300 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884232 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884261 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P28884297 |
- |
- |