Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P487918 |
BC046530.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P487918 |
BF147913.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P487918 |
AK168472.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P487918 |
BC044761.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P487918 |
BC044878.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P487918 |
BC046530.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P487918 |
BF147913.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P487918 |
AK168472.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P487918 |
BC044761.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P487918 |
BC044878.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431758 |
AF539600.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431758 |
AY211081.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
431758 |
NM_177634.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
683379 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
707092 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
256096 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
58684 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
902177 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P163010 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P487918 |
BC046530.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P487918 |
BF147913.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P487918 |
AK168472.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P487918 |
BC044761.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P487918 |
BC044878.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P487918 |
BC046530.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P487918 |
BF147913.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P487918 |
AK168472.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P487918 |
BC044761.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P487918 |
BC044878.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023580 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023575 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023577 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023624 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023595 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023606 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023582 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023612 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023617 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023602 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023610 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023593 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023591 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023619 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023600 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023609 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023601 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023604 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023618 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023613 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023611 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023608 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023586 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023578 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023574 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023621 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023598 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023589 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023596 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023599 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023584 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023616 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023581 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P25023622 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01490601 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P163010 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06150398 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P25548 |
BC046530.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P25548 |
AK168472.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P25548 |
BC044761.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P25548 |
BC044878.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P25548 |
AK030993.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P294566 |
AK020687.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P487918 |
BC046530.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P487918 |
BF147913.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P487918 |
AK168472.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P487918 |
BC044761.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P487918 |
BC044878.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P487918 |
BC046530.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P487918 |
BF147913.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P487918 |
AK168472.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P487918 |
BC044761.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P487918 |
BC044878.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P121883 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P134095 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P116738 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P01490601 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P163010 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06327315 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06327310 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06111935 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06111950 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06327324 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06117607 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06111940 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023630 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023628 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023597 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023594 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023598 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023635 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023591 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023654 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023616 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023642 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023600 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023587 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023578 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023575 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023609 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023631 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023652 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023604 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023574 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023606 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023577 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023589 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023592 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023586 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023608 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023612 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023638 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023625 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023614 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023580 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023622 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023651 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023599 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023611 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023639 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25023582 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06114395 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06579905 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023618 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023619 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06111950 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023606 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023582 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06327319 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023611 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023608 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05982321 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023602 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023614 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06574387 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023592 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023625 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023616 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023617 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023586 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023622 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023578 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023591 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023574 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023601 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023580 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023589 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023600 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023584 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023587 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023599 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023598 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023624 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06574388 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023604 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25023610 |
- |
- |