Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf2ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009970.2 | NP_034100.2 | 6 - 7 | 101 | ||
GenBank mRNA | BC150696.1 | - | 6 - 7 | 690 | 9087607 | 101 |
BU614102.1 | - | 4 - 5 | 101 | |||
M85078.1 | - | 6 - 7 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |