Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028762.1 | NP_083038.1 | 18 - 19 | 2378 | 66 | |
GenBank mRNA | AK004657.1 | - | 9 - 10 | 1193 | 66 | |
AK146493.1 | - | 18 - 19 | 2411 | 66 | ||
AK167100.1 | - | 18 - 19 | 2375 | 66 | ||
BC025619.1 | - | 18 - 19 | 2378 | 5320947 | 66 | |
BC034010.1 | - | 18 - 19 | 2344 | 4225239 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |