Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028639.3 | NP_082915.1 | 17 - 18 | 2355 | 65 | |
GenBank mRNA | AK040578.1 | - | 17 - 18 | 2358 | 65 | |
AK040610.1 | - | 17 - 18 | 2326 | 65 | ||
AK050263.1 | - | 17 - 18 | 2342 | 65 | ||
AK080284.1 | - | 17 - 18 | 2334 | 65 | ||
AK153704.1 | - | 17 - 18 | 2337 | 65 | ||
AK165244.1 | - | 10 - 11 | 1017 | 65 | ||
BC023773.1 | - | 13 - 14 | 1313 | 65 | ||
BC042512.1 | - | 17 - 18 | 2032 | 4223801 | 65 | |
BC049254.1 | - | 1 - 2 | 78 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |