Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ylpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178363.3 | NP_848140.3 | 8 - 9 | 5179 | 104 | |
GenBank mRNA | AB033168.1 | - | 8 - 9 | 2852 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |