Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039354.1 | NP_001034443.1 | 2 - 3 | 233 | 85 | |
GenBank mRNA | BC029721.1 | - | 2 - 3 | 139 | 85 | |
BG288381.1 | - | 2 - 3 | 171 | 85 | ||
BU923246.1 | - | 2 - 3 | 200 | 85 | ||
BY123635.1 | - | 2 - 3 | 233 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |