Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pof1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181579.1 | NP_853557.1 | 14 - 15 | 1678 | 100 | |
GenBank mRNA | AF309775.1 | - | 13 - 14 | 1571 | 100 | |
AK137653.1 | - | 14 - 15 | 1677 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |