Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem59l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182991.2 | NP_892036.1 | 3 - 4 | 428 | 87 | |
GenBank mRNA | AK005155.1 | - | 3 - 4 | 392 | 87 | |
BC056172.1 | - | 3 - 4 | 428 | 5699902 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |