Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025520.3 | NP_079796.1 | 1 - 2 | 78 | 75 | |
GenBank mRNA | AK008480.1 | - | 1 - 2 | 56 | 75 | |
AK009617.1 | - | 1 - 2 | 72 | 75 | ||
AK011218.1 | - | 1 - 2 | 64 | 75 | ||
BC048459.1 | - | 1 - 2 | 53 | 6601124 | 75 | |
BC061085.1 | - | 62 | 30295556 | 75 | ||
BC132626.1 | - | 1 - 2 | 78 | 40130903 | 75 | |
BC132628.1 | - | 1 - 2 | 78 | 40130905 | 75 | |
CB204385.1 | - | 1 - 2 | 71 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |