Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700012035.mm.1 |
NM_180974.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012035.mm.1 |
NM_180974.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012035.mm.1 |
TC1700001319.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012032.mm.1 |
NM_180974.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012032.mm.1 |
NM_180974.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012032.mm.1 |
TC1700001319.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012034.mm.1 |
NM_180974.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012034.mm.1 |
NM_180974.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012034.mm.1 |
TC1700001319.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012030.mm.1 |
NM_180974.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012030.mm.1 |
NM_180974.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012030.mm.1 |
TC1700001319.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012037.mm.1 |
NM_180974.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012037.mm.1 |
NM_180974.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012037.mm.1 |
TC1700001319.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700012036.mm.1 |
NM_180974.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012036.mm.1 |
NM_180974.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700012036.mm.1 |
TC1700001319.mm.1 |
MTA TC |