Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133800.3 | NP_598561.2 | 3 - 4 | 295 | 61 | |
GenBank mRNA | AK036891.1 | - | 3 - 4 | 238 | 61 | |
AK037035.1 | - | 3 - 4 | 289 | 61 | ||
AK083539.1 | - | 297 | 61 | |||
AK167446.1 | - | 3 - 4 | 289 | 61 | ||
AY940114.1 | - | 3 - 4 | 238 | 61 | ||
BC013701.1 | - | 3 - 4 | 280 | 3483364 | 61 | |
BC094424.1 | - | 3 - 4 | 282 | 6819926 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |