Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Patz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253691.1 | NP_001240620.1 | 2 - 3 | 1488 | 98 | |
NR_045108.2 | 2 - 3 | 277 | 98 | |||
GenBank mRNA | AK166189.1 | - | 2 - 3 | 1609 | 98 | |
BC055947.1 | - | 2 - 3 | 127 | 98 | ||
CA327687.1 | - | 2 - 3 | 77 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |