Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172477.4 | NP_766065.1 | 9 - 10 | 2260 | 86 | |
GenBank mRNA | AK048041.1 | - | 9 - 10 | 2245 | 86 | |
AK076686.1 | - | 1 - 2 | 403 | 86 | ||
AK081176.1 | - | 9 - 10 | 2261 | 86 | ||
AK160784.1 | - | 9 - 10 | 2022 | 86 | ||
AK165202.1 | - | 9 - 10 | 2115 | 86 | ||
AK167298.1 | - | 9 - 10 | 86 | |||
BC056367.1 | - | 9 - 10 | 2130 | 86 | ||
BC057310.1 | - | 9 - 10 | 2130 | 86 | ||
BC145599.1 | - | 8 - 9 | 1939 | 9054165 | 86 | |
BC150799.1 | - | 8 - 9 | 1939 | 9087710 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |