Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013491.2 | NP_038519.1 | 3 - 4 | 509 | 67 | |
GenBank mRNA | AK036735.1 | - | 3 - 4 | 511 | 67 | |
AY046403.1 | - | 3 - 4 | 334 | 67 | ||
AY046404.1 | - | 3 - 4 | 334 | 67 | ||
BC114336.1 | - | 3 - 4 | 474 | 40048966 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |