Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009054.3 | NP_033080.2 | 1 - 2 | 983 | 130 | |
GenBank mRNA | AK031744.1 | - | 1 - 2 | 701 | 130 | |
AK088180.1 | - | 1 - 2 | 467 | 130 | ||
BC003219.1 | - | 1 - 2 | 488 | 3482042 | 130 | |
BC069924.1 | - | 1 - 2 | 694 | 130 | ||
BC085503.1 | - | 1 - 2 | 690 | 130 | ||
L46855.1 | - | 594 | 130 | |||
X75343.1 | - | 1 - 2 | 398 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |