Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500001782.mm.1 |
NM_026340.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001782.mm.1 |
NM_026340.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001782.mm.1 |
NM_152894.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001782.mm.1 |
NM_152894.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001782.mm.1 |
TC1500000240.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500001783.mm.1 |
NM_026340.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001783.mm.1 |
NM_026340.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001783.mm.1 |
NM_152894.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001783.mm.1 |
NM_152894.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001783.mm.1 |
TC1500000240.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500001784.mm.1 |
NM_026340.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001784.mm.1 |
NM_026340.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001784.mm.1 |
NM_152894.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001784.mm.1 |
NM_152894.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001784.mm.1 |
TC1500000240.mm.1 |
MTA TC |