Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013782.4 | NP_038810.2 | 5 - 6 | 618 | 69 | |
GenBank mRNA | AF099053.1 | - | 5 - 6 | 502 | 69 | |
AK021249.1 | - | 5 - 6 | 583 | 69 | ||
AK031906.1 | - | 5 - 6 | 604 | 69 | ||
AK032686.1 | - | 5 - 6 | 583 | 69 | ||
AK036368.1 | - | 5 - 6 | 619 | 69 | ||
AK048026.1 | - | 5 - 6 | 601 | 69 | ||
AK081275.1 | - | 5 - 6 | 585 | 69 | ||
AK137415.1 | - | 578 | 69 | |||
BC009013.1 | - | 3 - 4 | 285 | 69 | ||
BC053517.1 | - | 5 - 6 | 555 | 6315175 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |