Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033324.2 | NP_201581.2 | 4 - 5 | 1370 | 106 | |
GenBank mRNA | AB086857.1 | - | 4 - 5 | 1370 | 106 | |
AK048056.1 | - | 3 - 4 | 1171 | 106 | ||
BC051013.1 | - | 2 - 3 | 1556 | 106 | ||
BC062919.1 | - | 4 - 5 | 1390 | 6833280 | 106 | |
BC066118.1 | - | 4 - 5 | 1417 | 6400561 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |