Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_028401.1 | 9 - 10 | 1096 | 146 | ||
GenBank mRNA | AK012656.1 | - | 7 - 8 | 945 | 146 | |
AK014920.1 | - | 9 - 10 | 1048 | 146 | ||
AK050165.1 | - | 9 - 10 | 1052 | 146 | ||
AK166329.1 | - | 9 - 10 | 1051 | 146 | ||
AK166633.1 | - | 8 - 9 | 2166 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |