Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011915.2 | NP_036045.1 | 1 - 2 | 482 | 68 | |
GenBank mRNA | AF122923.1 | - | 1 - 2 | 256 | 68 | |
AK077698.1 | - | 1 - 2 | 483 | 68 | ||
AK142443.1 | - | 1 - 2 | 483 | 68 | ||
AK158359.1 | - | 1 - 2 | 483 | 68 | ||
BC013268.1 | - | 1 - 2 | 283 | 3984128 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |