Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178414.3 | NP_848501.1 | 1 - 2 | 145 | 97 | |
GenBank mRNA | AK132127.1 | - | 1 - 2 | 146 | 97 | |
BC048390.1 | - | 1 - 2 | 81 | 6312894 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |