Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145469.5 | NP_663444.2 | 5 - 6 | 569 | 119 | |
GenBank mRNA | AK033975.1 | - | 5 - 6 | 570 | 119 | |
BC016107.1 | - | 5 - 6 | 592 | 4921508 | 119 | |
BC030399.1 | - | 5 - 6 | 1086 | 4166825 | 119 | |
BC058207.1 | - | 5 - 6 | 735 | 5361417 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |