Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163024.1 | NP_001156496.1 | 2 - 3 | 718 | 93 | |
NM_001163025.1 | NP_001156497.1 | 2 - 3 | 718 | 93 | ||
NM_153395.2 | NP_700444.2 | 2 - 3 | 718 | 93 | ||
GenBank mRNA | AA542162.1 | - | 2 - 3 | 206 | 93 | |
AB014059.1 | - | 2 - 3 | 240 | 93 | ||
AK122425.1 | - | 2 - 3 | 718 | 93 | ||
AK161217.1 | - | 2 - 3 | 597 | 93 | ||
BC064131.1 | - | 2 - 3 | 208 | 93 | ||
BC150943.1 | - | 2 - 3 | 239 | 9087854 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |