Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Athl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK171694.1 | - | 1 - 2 | 255 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |