Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198606.2 | NP_941008.2 | 4 - 5 | 580 | 89 | |
GenBank mRNA | BC060375.1 | - | 4 - 5 | 522 | 5038466 | 89 |
BU518555.1 | - | 4 - 5 | 538 | 89 | ||
CB950393.1 | - | 4 - 5 | 584 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |