Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178758.3 | NP_848873.1 | 4 - 5 | 809 | 88 | |
GenBank mRNA | AK040650.1 | - | 4 - 5 | 746 | 88 | |
AK050132.1 | - | 5 - 6 | 906 | 88 | ||
AK050219.1 | - | 4 - 5 | 834 | 88 | ||
AK050288.1 | - | 4 - 5 | 829 | 88 | ||
AK050404.1 | - | 4 - 5 | 832 | 88 | ||
AK050458.1 | - | 4 - 5 | 771 | 88 | ||
AK080219.1 | - | 4 - 5 | 745 | 88 | ||
AK149591.1 | - | 4 - 5 | 829 | 88 | ||
BC061229.1 | - | 4 - 5 | 733 | 88 | ||
BC095985.1 | - | 4 - 5 | 809 | 4971395 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |