Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171003.1 | NP_001164474.1 | 45 - 46 | 5361 | 65 | |
GenBank mRNA | AK031332.1 | - | 29 - 30 | 3364 | 65 | |
AK033548.1 | - | 5 - 6 | 500 | 65 | ||
BC025509.1 | - | 11 - 12 | 1061 | 65 | ||
EU937529.1 | - | 45 - 46 | 5361 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |