Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfatc2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010900.3 | NP_035030.2 | 4 - 5 | 660 | 74 | |
GenBank mRNA | AK005947.1 | - | 4 - 5 | 651 | 74 | |
BC113761.1 | - | 4 - 5 | 661 | 40054445 | 74 | |
U76759.1 | - | 4 - 5 | 650 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |