Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009513.2 | NP_033539.1 | 2 - 3 | 224 | 63 | |
GenBank mRNA | AK138191.1 | - | 2 - 3 | 155 | 63 | |
AK160941.1 | - | 2 - 3 | 209 | 63 | ||
AU080650.1 | - | 2 - 3 | 310 | 63 | ||
BC052019.1 | - | 1 - 2 | 47 | 5708003 | 63 | |
D83206.1 | - | 2 - 3 | 177 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |