Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276452.1 | NP_001263381.1 | 8 - 9 | 1106 | 126 | |
NM_172773.3 | NP_766361.1 | 8 - 9 | 1184 | 126 | ||
GenBank mRNA | AK029102.1 | - | 8 - 9 | 1167 | 126 | |
AK169868.1 | - | 8 - 9 | 1133 | 126 | ||
BC058785.1 | - | 8 - 9 | 1076 | 6309785 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |