Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lppr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290299.1 | NP_001277228.1 | 2 - 3 | 320 | 100 | |
NM_144935.1 | NP_659184.1 | 1 - 2 | 292 | 100 | ||
GenBank mRNA | AK082453.1 | - | 2 - 3 | 283 | 100 | |
AY438023.1 | - | 1 - 2 | 292 | 100 | ||
BC018242.1 | - | 1 - 2 | 292 | 4503683 | 100 | |
BC023082.1 | - | 2 - 3 | 297 | 5357160 | 100 | |
CX122376.1 | - | 2 - 3 | 320 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |