Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700005188.mm.1 |
TC0700000575.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700005188.mm.1 |
NM_001166173.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005188.mm.1 |
NM_001166173.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005188.mm.1 |
NM_001166174.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005188.mm.1 |
NM_001166174.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005188.mm.1 |
NM_028618.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005188.mm.1 |
NM_028618.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005188.mm.1 |
NM_172899.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005188.mm.1 |
NM_172899.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005189.mm.1 |
TC0700000575.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700005189.mm.1 |
NM_001166173.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005189.mm.1 |
NM_001166173.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005189.mm.1 |
NM_001166174.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005189.mm.1 |
NM_001166174.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005189.mm.1 |
NM_028618.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005189.mm.1 |
NM_028618.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005189.mm.1 |
NM_172899.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005189.mm.1 |
NM_172899.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005190.mm.1 |
TC0700000575.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700005190.mm.1 |
NM_001166173.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005190.mm.1 |
NM_001166173.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005190.mm.1 |
NM_001166174.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005190.mm.1 |
NM_001166174.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005190.mm.1 |
NM_028618.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005190.mm.1 |
NM_028618.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005190.mm.1 |
NM_172899.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005190.mm.1 |
NM_172899.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005186.mm.1 |
TC0700000575.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700005186.mm.1 |
NM_028618.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700005186.mm.1 |
NM_028618.2 |
Refseq |