Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081114.1 | NP_001074583.1 | 9 - 10 | 1332 | 98 | |
GenBank mRNA | AK005167.1 | - | 1 - 2 | 129 | 98 | |
BC056173.1 | - | 1 - 2 | 100 | 98 | ||
BC138413.1 | - | 8 - 9 | 1251 | 8861433 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |