Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vsig2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020518.2 | NP_065264.2 | 5 - 6 | 827 | 57 | |
GenBank mRNA | AK004478.1 | - | 4 - 5 | 547 | 57 | |
AK008920.1 | - | 5 - 6 | 789 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |