Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008168.2 | NP_032194.2 | 1 - 2 | 332 | 113 | |
GenBank mRNA | BC052009.1 | - | 1 - 2 | 220 | 5695215 | 113 |
BC058097.1 | - | 1 - 2 | 66 | 113 | ||
BC110682.1 | - | 1 - 2 | 255 | 5033584 | 113 | |
BU701817.1 | - | 1 - 2 | 332 | 113 | ||
CJ143126.1 | - | 1 - 2 | 308 | 113 | ||
D10011.1 | - | 1 - 2 | 261 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |