Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008168.2 | NP_032194.2 | 6 - 7 | 1069 | 65 | |
GenBank mRNA | BC052009.1 | - | 6 - 7 | 957 | 5695215 | 65 |
BC058097.1 | - | 6 - 7 | 828 | 65 | ||
BC110682.1 | - | 6 - 7 | 992 | 5033584 | 65 | |
D10011.1 | - | 6 - 7 | 998 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |