Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lactb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030717.1 | NP_109642.1 | 1 - 2 | 444 | 73 | |
GenBank mRNA | AF317900.1 | - | 1 - 2 | 444 | 73 | |
AK078550.1 | - | 1 - 2 | 382 | 73 | ||
AK087297.1 | - | 1 - 2 | 412 | 73 | ||
AK149410.1 | - | 1 - 2 | 387 | 73 | ||
BC021604.1 | - | 1 - 2 | 370 | 73 | ||
BC147809.1 | - | 1 - 2 | 389 | 9087526 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |