Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pml | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178087.4 | NP_835188.2 | 5 - 6 | 1524 | 69 | |
GenBank mRNA | AK028044.1 | - | 5 - 6 | 1526 | 69 | |
AK032799.1 | - | 3 - 4 | 404 | 69 | ||
AK137538.1 | - | 5 - 6 | 1472 | 69 | ||
KJ650238.1 | - | 5 - 6 | 1428 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |