Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008297.3 | NP_032323.3 | 4 - 5 | 582 | 78 | |
GenBank mRNA | AK052270.1 | - | 4 - 5 | 481 | 78 | |
AK053976.1 | - | 4 - 5 | 583 | 78 | ||
BC018414.1 | - | 4 - 5 | 504 | 3990765 | 78 | |
X61754.1 | - | 4 - 5 | 616 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |