Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtif2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282118.1 | NP_001269047.1 | 5 - 6 | 846 | 83 | |
NM_001282119.1 | NP_001269048.1 | 5 - 6 | 768 | 83 | ||
NM_001282120.1 | NP_001269049.1 | 5 - 6 | 1210 | 83 | ||
NM_133767.3 | NP_598528.2 | 5 - 6 | 1124 | 83 | ||
GenBank mRNA | AK010766.1 | - | 5 - 6 | 1069 | 83 | |
AK136344.1 | - | 5 - 6 | 808 | 83 | ||
AK146961.1 | - | 5 - 6 | 1064 | 83 | ||
BC016590.1 | - | 5 - 6 | 700 | 3585359 | 83 | |
BC066819.1 | - | 5 - 6 | 1104 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |