Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133872.2 | NP_598633.2 | 4 - 5 | 873 | 79 | |
GenBank mRNA | AK129170.1 | - | 4 - 5 | 813 | 79 | |
BC059885.1 | - | 4 - 5 | 860 | 6815975 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |