Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pafah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285872.1 | NP_001272801.1 | 1 - 2 | 120 | 106 | |
GenBank mRNA | BC025495.1 | - | 1 - 2 | 131 | 5310200 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |