Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prickle2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081146.2 | NP_001074615.1 | 5 - 6 | 601 | 67 | |
NM_001134459.1 | NP_001127931.1 | 4 - 5 | 680 | 67 | ||
NM_001134460.2 | NP_001127932.1 | 4 - 5 | 1002 | 67 | ||
NM_001134461.1 | NP_001127933.1 | 4 - 5 | 497 | 67 | ||
GenBank mRNA | AK147576.1 | - | 4 - 5 | 681 | 67 | |
AK147628.1 | - | 4 - 5 | 681 | 67 | ||
BC050793.1 | - | 4 - 5 | 497 | 67 | ||
BC099897.1 | - | 5 - 6 | 572 | 30621797 | 67 | |
BC145754.1 | - | 4 - 5 | 519 | 40131057 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |