Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thumpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008188.2 | NP_032214.1 | 4 - 5 | 905 | 82 | |
GenBank mRNA | AK142140.1 | - | 4 - 5 | 924 | 82 | |
BC002024.1 | - | 4 - 5 | 907 | 3591186 | 82 | |
BC012688.1 | - | 3 - 4 | 853 | 3987438 | 82 | |
BC046800.1 | - | 4 - 5 | 905 | 6403880 | 82 | |
U83176.1 | - | 4 - 5 | 899 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |